5-Acetamidofluorescein di-(β-D-galactopyranoside) ≥95%
Dodavatel: Apollo Scientific
Synonyms:
C2FDG
BICL4232-10MG
BICL4232-2MG
BICL4232-5MG
APOSBICL4232-10MGEA
29760
CZK
APOSBICL4232-10MG
APOSBICL4232-2MG
APOSBICL4232-5MG
5-Acetamidofluorescein di-(β-D-galactopyranoside) ≥95%
5-Acetamidofluorescein di-(β-D-galactopyranoside)
Vzorec:
C₃₄H₃₅NO₁₆ MW: 713.639 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00467511 CAS číslo: 216299-45-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...