Tisk…

2-Nitrophenyl α-D-galactopyranoside ≥98%

Dodavatel: Apollo Scientific
Varování

BICL4229-100MG BICL4229-250MG BICL4229-500MG
APOSBICL4229-100MGEA 10350 CZK
APOSBICL4229-100MG APOSBICL4229-250MG APOSBICL4229-500MG
2-Nitrophenyl α-D-galactopyranoside ≥98%
2-Nitrophenyl α-D-galactopyranoside
Vzorec: C₁₂H₁₅NO₈
MW: 301,249 g/mol
Bod tání: 150…151 °C
Storage Temperature: Freezer
MDL Number: MFCD00069785
CAS číslo: 19887-85-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR