Acetaminophen β-D-glucuronide ≥95%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-Acetamidophenyl-O-β-D-glucuronide, Paracetamol-O-β-D-glucuronide, p-Acetylaminophenol-O-β-D-glucuronide
BICL4227-10MG
BICL4227-25MG
BICL4227-50MG
APOSBICL4227-10MGEA
7910
CZK
APOSBICL4227-10MG
APOSBICL4227-25MG
APOSBICL4227-50MG
Acetaminophen β-D-glucuronide ≥95%
Acetaminophen β-D-glucuronide
Vzorec:
C₁₄H₁₇NO₈ MW: 327,287 g/mol Bod tání: 185…190 °C (rozkl.) Storage Temperature: Refrigerator |
MDL Number:
MFCD03094680 CAS číslo: 16110-10-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...