Tisk…

2,3,4-Tri-O-acetyl-1,6-diazido-1,6-dideoxy-β-D-glucopyranoside ≥98%

Dodavatel: Apollo Scientific

Synonyms: 2,3,4-Tri-O-acetyl-6-azido-6-deoxy-β-D-glucopyranosyl azide

BICL4213-250MG BICL4213-500MG BICL4213-100MG
APOSBICL4213-250MGEA 17920 CZK
APOSBICL4213-250MG APOSBICL4213-500MG APOSBICL4213-100MG
2,3,4-Tri-O-acetyl-1,6-diazido-1,6-dideoxy-β-D-glucopyranoside ≥98%
2,3,4-Tri-O-acetyl-1,6-diazido-1,6-dideoxy-β-D-glucopyranoside
Storage Temperature: Ambient
CAS číslo: 106192-56-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR