Tisk…

6-O-Trityl-D-glucopyranose ≥99%

Dodavatel: Apollo Scientific

Synonyms: 6-O-Triphenylmethyl-D-glucopyranose

BICL4206-1G BICL4206-250MG
APOSBICL4206-1GEA 10650 CZK
APOSBICL4206-1G APOSBICL4206-250MG
6-O-Trityl-D-glucopyranose ≥99%
6-O-Trityl-D-glucopyranose
Storage Temperature: Ambient
CAS číslo: 67919-34-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR