Tisk…

1,2,4,6-Tetra-O-acetyl-3-O-benzyl-β-D-glucopyranose ≥99%

Dodavatel: Apollo Scientific

BICL4205-2G BICL4205-5G BICL4205-1G
APOSBICL4205-2GEA 12730 CZK
APOSBICL4205-2G APOSBICL4205-5G APOSBICL4205-1G
1,2,4,6-Tetra-O-acetyl-3-O-benzyl-β-D-glucopyranose ≥99%
1,2,4,6-Tetra-O-acetyl-3-O-benzyl-β-D-glucopyranose
Storage Temperature: Ambient
CAS číslo: 39686-94-7

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR