Tarenflurbil-acyl-β-D-glucuronide ≥97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
(R)-Flurbiprofen-acyl-β-D-glucuronide
BICL4202-1MG
BICL4202-5MG
BICL4202-2MG
APOSBICL4202-1MGEA
11540
CZK
APOSBICL4202-1MG
APOSBICL4202-5MG
APOSBICL4202-2MG
Tarenflurbil-acyl-β-D-glucuronide ≥97%
Tarenflurbil-acyl-β-D-glucuronide
Storage Temperature:
Freezer |
CAS číslo:
162992-67-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...