Telmisartan-acyl-β-D-glucuronide ≥95%
Dodavatel: Apollo Scientific
BICL4201-5MG
BICL4201-1MG
BICL4201-2MG
APOSBICL4201-5MGEA
26250
CZK
APOSBICL4201-5MG
APOSBICL4201-1MG
APOSBICL4201-2MG
Telmisartan-acyl-β-D-glucuronide ≥95%
Telmisartan-acyl-β-D-glucuronide
Storage Temperature:
Ambient |
CAS číslo:
250780-40-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...