Repaglinide-acyl-β-D-glucuronide ≥95%
Dodavatel: Apollo Scientific
BICL4193-1MG
BICL4193-5MG
BICL4193-2MG
APOSBICL4193-1MGEA
10750
CZK
APOSBICL4193-1MG
APOSBICL4193-5MG
APOSBICL4193-2MG
Repaglinide-acyl-β-D-glucuronide ≥95%
Repaglinide-acyl-β-D-glucuronide
Storage Temperature:
Ambient |
CAS číslo:
1309112-13-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...