Tisk…

2-Nitrophenyl β-D-glucuronide ≥99%

Dodavatel: Apollo Scientific

BICL4183-100MG BICL4183-250MG BICL4183-500MG
APOSBICL4183-100MGEA 11970 CZK
APOSBICL4183-100MG APOSBICL4183-250MG APOSBICL4183-500MG
2-Nitrophenyl β-D-glucuronide ≥99%
2-Nitrophenyl β-D-glucuronide
Storage Temperature: Ambient
CAS číslo: 137629-36-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR