Naringenin-7-O-β-D-glucuronide ≥98%
Dodavatel: Apollo Scientific
BICL4178-1MG
BICL4178-5MG
BICL4178-2MG
APOSBICL4178-1MGEA
8010
CZK
APOSBICL4178-1MG
APOSBICL4178-5MG
APOSBICL4178-2MG
Naringenin-7-O-β-D-glucuronide ≥98%
Naringenin-7-O-β-D-glucuronide
Storage Temperature:
Ambient |
CAS číslo:
158196-34-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...