Tisk…

Methyl 3-amino-3-deoxy-α-D-mannopyranoside hydrochloride ≥98%

Dodavatel: Apollo Scientific

BICL4174-1G BICL4174-500MG BICL4174-2G
APOSBICL4174-1GEA 15150 CZK
APOSBICL4174-1G APOSBICL4174-500MG APOSBICL4174-2G
Methyl 3-amino-3-deoxy-α-D-mannopyranoside hydrochloride ≥98%
Methyl 3-amino-3-deoxy-α-D-mannopyranoside hydrochloride
Storage Temperature: Ambient
CAS číslo: 14133-35-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR