Tisk…
Methyl 4,6-O-benzylidene-β-D-glucopyranoside ≥98%
Dodavatel: Apollo Scientific
BICL4161-10G BICL4161-25G BICL4161-5G
APOSBICL4161-10GEA 15940 CZK
APOSBICL4161-10G APOSBICL4161-25G APOSBICL4161-5G
Methyl 4,6-O-benzylidene-β-D-glucopyranoside ≥98%
Methyl 4,6-O-benzylidene-β-D-glucopyranoside
Storage Temperature: Ambient
CAS číslo: 14155-23-8