Tisk…

Methyl 6-O-acetyl-3-O-benzyl-2-benzyloxycarbonylamino-2-deoxy-4-O-(methyl 2-O-acetyl-3-O-benzyl-α-L-idopyranosyluronate)-α-D-glucopyranoside ≥99%

Dodavatel: Apollo Scientific

Synonyms: Methyl 6-O-acetyl-3-O-benzyl-N-Cbz-4-O-(methyl 2-O-acetyl-3-O-benzyl-α-L-idopyranosyluronate)-α-D-glucosaminide

BICL4160-25MG BICL4160-50MG
APOSBICL4160-25MGEA 23840 CZK
APOSBICL4160-25MG APOSBICL4160-50MG
Methyl 6-O-acetyl-3-O-benzyl-2-benzyloxycarbonylamino-2-deoxy-4-O-(methyl 2-O-acetyl-3-O-benzyl-α-L-idopyranosyluronate)-α-D-glucopyranoside ≥99%
Methyl 6-O-acetyl-3-O-benzyl-2-benzyloxycarbonylamino-2-deoxy-4-O-(methyl 2-O-acetyl-3-O-benzyl-α-L-idopyranosyluronate)-α-D-glucopyranoside
Storage Temperature: Ambient
CAS číslo: 114869-97-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR