Tisk…
Methyl 3-O-benzyl-2-benzyloxycarbonylamino-2-deoxy-α-D-glucopyranoside ≥99%
Dodavatel: Apollo Scientific
Synonyms: Methyl 3-O-benzyl-N-Cbz-α-D-glucosaminide
BICL4158-100MG BICL4158-250MG BICL4158-500MG
APOSBICL4158-100MGEA 8010 CZK
APOSBICL4158-100MG APOSBICL4158-250MG APOSBICL4158-500MG
Methyl 3-O-benzyl-2-benzyloxycarbonylamino-2-deoxy-α-D-glucopyranoside ≥99%
Methyl 3-O-benzyl-2-benzyloxycarbonylamino-2-deoxy-α-D-glucopyranoside
Storage Temperature: Ambient
CAS číslo: 87907-35-5