Tisk…

Methyl 2-benzyloxycarbonylamino-2-deoxy-α-D-glucopyranoside ≥98%

Dodavatel: Apollo Scientific

BICL4155-1G BICL4155-500MG BICL4155-250MG
APOSBICL4155-1GEA 27870 CZK
APOSBICL4155-1G APOSBICL4155-500MG APOSBICL4155-250MG
Methyl 2-benzyloxycarbonylamino-2-deoxy-α-D-glucopyranoside ≥98%
Methyl 2-benzyloxycarbonylamino-2-deoxy-α-D-glucopyranoside
Storage Temperature: Ambient
CAS číslo: 4704-15-8

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR