Tisk…

4-Methylumbelliferyl β-D-cellopentaoside ≥95%

Dodavatel: Apollo Scientific

Synonyms: 4-MU β-D-cellopentaoside

BICL4152-1MG BICL4152-2MG
APOSBICL4152-1MGEA 17530 CZK
APOSBICL4152-1MG APOSBICL4152-2MG
4-Methylumbelliferyl β-D-cellopentaoside ≥95%
4-Methylumbelliferyl β-D-cellopentaoside
Storage Temperature: Ambient
CAS číslo: 84325-20-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR