Tisk…

4-Methylumbelliferyl β-D-cellotetraoside ≥98%

Dodavatel: Apollo Scientific

Synonyms: 4-MU β-D-cellotetraoside

BICL4151-1MG BICL4151-2MG
APOSBICL4151-1MGEA 16340 CZK
APOSBICL4151-1MG APOSBICL4151-2MG
4-Methylumbelliferyl β-D-cellotetraoside ≥98%
4-Methylumbelliferyl β-D-cellotetraoside
Storage Temperature: Ambient
CAS číslo: 84325-19-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR