4-Methylumbelliferyl β-D-cellotrioside ≥98%
Dodavatel: Apollo Scientific
BICL4150-1MG
BICL4150-2MG
APOSBICL4150-1MGEA
13530
CZK
APOSBICL4150-1MG
APOSBICL4150-2MG
4-Methylumbelliferyl β-D-cellotrioside ≥98%
4-Methylumbelliferyl β-D-cellotrioside
Storage Temperature:
Ambient |
CAS číslo:
84325-18-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...