Methyl (4-methylphenyl 2,3,4-tri-O-acetyl-1-thio-β-D-glucopyranosid)uronate ≥99%
Dodavatel: Apollo Scientific
BICL4149-1G
BICL4149-250MG
APOSBICL4149-1GEA
60840
CZK
APOSBICL4149-1G
APOSBICL4149-250MG
Methyl (4-methylphenyl 2,3,4-tri-O-acetyl-1-thio-β-D-glucopyranosid)uronate ≥99%
Methyl (4-methylphenyl 2,3,4-tri-O-acetyl-1-thio-β-D-glucopyranosid)uronate
Vzorec:
C₂₀H₂₄O₉S Storage Temperature: Ambient |
MDL Number:
MFCD18643077 CAS číslo: 61025-09-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...