Tisk…

4-Methylphenyl 2,3,4-tri-O-benzyl-1-thio-β-D-glucopyranoside ≥98%

Dodavatel: Apollo Scientific

Synonyms: Tolyl 2,3,4-tri-O-benzyl-1-thio-β-D-glucopyranoside, Thiocresyl 2,3,4-tri-O-benzyl-β-D-glucopyranoside

BICL4147-500MG BICL4147-1G BICL4147-250MG
APOSBICL4147-500MGEA 18680 CZK
APOSBICL4147-500MG APOSBICL4147-1G APOSBICL4147-250MG
4-Methylphenyl 2,3,4-tri-O-benzyl-1-thio-β-D-glucopyranoside ≥98%
4-Methylphenyl 2,3,4-tri-O-benzyl-1-thio-β-D-glucopyranoside
Storage Temperature: Ambient
CAS číslo: 479252-05-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR