Tisk…

4-Methoxyphenyl 2,3,4-tri-O-benzyl-β-D-glucopyranoside ≥99%

Dodavatel: Apollo Scientific

BICL4137-1G BICL4137-2G BICL4137-5G
APOSBICL4137-1GEA 8010 CZK
APOSBICL4137-1G APOSBICL4137-2G APOSBICL4137-5G
4-Methoxyphenyl 2,3,4-tri-O-benzyl-β-D-glucopyranoside ≥99%
4-Methoxyphenyl 2,3,4-tri-O-benzyl-β-D-glucopyranoside
Storage Temperature: Ambient
CAS číslo: 1055325-21-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR