Tisk…

Mycophenolic acid acyl-β-D-glucuronide ≥90%

Dodavatel: Apollo Scientific
Varování

BICL4132-1MG BICL4132-5MG BICL4132-2MG
APOSBICL4132-1MGEA 11180 CZK
APOSBICL4132-1MG APOSBICL4132-5MG APOSBICL4132-2MG
Mycophenolic acid acyl-β-D-glucuronide ≥90%
Mycophenolic acid acyl-β-D-glucuronide
Vzorec: C₂₃H₂₈O₁₂
Bod varu: 805 °C
Bod tání: 90 °C
Hustota: 1,5 g/cm³ (25 °C)
Storage Temperature: Freezer
MDL Number: MFCD11977846
CAS číslo: 99043-04-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR