Tisk…
Mycophenolic acid-O-β-D-glucuronide ≥97%
Dodavatel: Apollo Scientific
Synonyms: Mycophenolic acid glucuronide, Mycophenolic acid-phenolic-β-D-glucuronide, (2S,3S,4S,5R,6S)-6-((5-((E)-5-Carboxy-3-methylpent-2-en-1-yl)-6-methoxy-7-methyl-3-oxo-1,3-dihydroisobenzofuran-4-yl)oxy)-3,4,5-trihydroxytetrahydro-2H-pyran-2-carboxylic acid
BICL4131-1MG BICL4131-5MG BICL4131-2MG
APOSBICL4131-1MGEA 7210 CZK
APOSBICL4131-1MG APOSBICL4131-5MG APOSBICL4131-2MG
Mycophenolic acid-O-β-D-glucuronide ≥97%
Mycophenolic acid-O-β-D-glucuronide
Vzorec: C₂₃H₂₈O₁₂
MW: 496.461 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09878308
CAS číslo: 31528-44-6