Tisk…

(R,S)-Ibuprofen-acyl-β-D-glucuronide ≥98%

Dodavatel: Apollo Scientific

BICL4117-1MG BICL4117-5MG BICL4117-2MG
APOSBICL4117-1MGEA 9200 CZK
APOSBICL4117-1MG APOSBICL4117-5MG APOSBICL4117-2MG
(R,S)-Ibuprofen-acyl-β-D-glucuronide ≥98%
(R,S)-Ibuprofen-acyl-β-D-glucuronide
Storage Temperature: Ambient
CAS číslo: 115075-59-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR