Tisk…

Isosorbide-5-mononitrate-2-O-β-D-glucuronide ≥97%

Dodavatel: Apollo Scientific

BICL4114-1MG BICL4114-5MG BICL4114-2MG
APOSBICL4114-1MGEA 11180 CZK
APOSBICL4114-1MG APOSBICL4114-5MG APOSBICL4114-2MG
Isosorbide-5-mononitrate-2-O-β-D-glucuronide ≥97%
Isosorbide-5-mononitrate-2-O-β-D-glucuronide
Storage Temperature: Ambient
CAS číslo: 32871-20-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR