Indomethacin-acyl-β-D-glucuronide ≥95%
Dodavatel: Apollo Scientific
Synonyms:
Indometacin-acyl-β-D-glucuronide
BICL4112-1MG
BICL4112-2MG
APOSBICL4112-1MGEA
14350
CZK
APOSBICL4112-1MG
APOSBICL4112-2MG
Indomethacin-acyl-β-D-glucuronide ≥95%
Indomethacin-acyl-β-D-glucuronide
Storage Temperature:
Ambient |
CAS číslo:
75523-11-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...