Tisk…
Gemfibrozil-acyl-β-D-glucuronide ≥95%
Dodavatel: Apollo Scientific
Varování
BICL4101-1MG BICL4101-2MG
APOSBICL4101-1MGEA 12730 CZK
APOSBICL4101-1MG APOSBICL4101-2MG
Gemfibrozil-acyl-β-D-glucuronide ≥95%
Gemfibrozil-acyl-β-D-glucuronide
Storage Temperature: Freezer
CAS číslo: 91683-38-4