Tisk…

(S)-Flurbiprofen-acyl-β-D-glucuronide ≥98%

Dodavatel: Apollo Scientific

BICL4099-1MG BICL4099-2MG
APOSBICL4099-1MGEA 12370 CZK
APOSBICL4099-1MG APOSBICL4099-2MG
(S)-Flurbiprofen-acyl-β-D-glucuronide ≥98%
(S)-Flurbiprofen-acyl-β-D-glucuronide
Storage Temperature: Ambient
CAS číslo: 162992-66-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR