Etodolac-acyl-β-D-glucuronide ≥95%
Dodavatel: Apollo Scientific
BICL4092-1MG
BICL4092-5MG
BICL4092-2MG
APOSBICL4092-1MGEA
10750
CZK
APOSBICL4092-1MG
APOSBICL4092-5MG
APOSBICL4092-2MG
Etodolac-acyl-β-D-glucuronide ≥95%
Etodolac-acyl-β-D-glucuronide
Storage Temperature:
Ambient |
CAS číslo:
79541-43-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...