3,5-Di-O-benzoyl-1,2-O-isopropylidene-α-D-ribofuranose ≥99%
Dodavatel: Apollo Scientific
BICL4087-250MG
BICL4087-500MG
BICL4087-100MG
APOSBICL4087-250MGEA
13530
CZK
APOSBICL4087-250MG
APOSBICL4087-500MG
APOSBICL4087-100MG
3,5-Di-O-benzoyl-1,2-O-isopropylidene-α-D-ribofuranose ≥99%
3,5-Di-O-benzoyl-1,2-O-isopropylidene-α-D-ribofuranose
Storage Temperature:
Ambient |
CAS číslo:
80244-95-7 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...