3,4-Di-O-acetyl-1,6-anhydro-2-deoxy-2-fluoro-β-D-glucopyranose ≥99%
Dodavatel: Apollo Scientific
Synonyms:
3,4-Di-O-acetyl-2-fluoro-β-D-levoglucosan
BICL4074-100MG
BICL4074-250MG
BICL4074-500MG
APOSBICL4074-100MGEA
9200
CZK
APOSBICL4074-100MG
APOSBICL4074-250MG
APOSBICL4074-500MG
3,4-Di-O-acetyl-1,6-anhydro-2-deoxy-2-fluoro-β-D-glucopyranose ≥99%
3,4-Di-O-acetyl-1,6-anhydro-2-deoxy-2-fluoro-β-D-glucopyranose
Storage Temperature:
Ambient |
CAS číslo:
23236-00-2 |