Tisk…

1,6-Di-O-acetyl-2-azido-3,4-di-O-benzyl-2-deoxy-α-D-glucopyranose ≥99%

Dodavatel: Apollo Scientific

BICL4071-1G BICL4071-500MG BICL4071-250MG
APOSBICL4071-1GEA 38020 CZK
APOSBICL4071-1G APOSBICL4071-500MG APOSBICL4071-250MG
1,6-Di-O-acetyl-2-azido-3,4-di-O-benzyl-2-deoxy-α-D-glucopyranose ≥99%
1,6-Di-O-acetyl-2-azido-3,4-di-O-benzyl-2-deoxy-α-D-glucopyranose
Storage Temperature: Ambient
CAS číslo: 55682-49-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR