3,4-Di-O-acetyl-1,6-anhydro-2-deoxy-2-iodo-β-D-glucopyranose ≥99%
Dodavatel: Apollo Scientific
Synonyms:
3,4-Di-O-acetyl-2-iodo-β-D-levoglucosan
BICL4070-1G
BICL4070-500MG
APOSBICL4070-1GEA
43970
CZK
APOSBICL4070-1G
APOSBICL4070-500MG
3,4-Di-O-acetyl-1,6-anhydro-2-deoxy-2-iodo-β-D-glucopyranose ≥99%
3,4-Di-O-acetyl-1,6-anhydro-2-deoxy-2-iodo-β-D-glucopyranose
Storage Temperature:
Ambient |
CAS číslo:
136573-62-1 |