Tisk…

Dexibuprofen-acyl-β-D-glucuronide ≥95%

Dodavatel: Apollo Scientific

Synonyms: (S)-Ibuprofen-acyl-β-D-glucuronide

BICL4061-1MG BICL4061-5MG BICL4061-2MG
APOSBICL4061-1MGEA 11540 CZK
APOSBICL4061-1MG APOSBICL4061-5MG APOSBICL4061-2MG
Dexibuprofen-acyl-β-D-glucuronide ≥95%
Dexibuprofen-acyl-β-D-glucuronide
Storage Temperature: Ambient
CAS číslo: 98649-76-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR