Tisk…

2-Chloro-4-nitrophenyl α-D-glucopyranoside ≥98%

Dodavatel: Apollo Scientific

BICL4056-100MG BICL4056-250MG BICL4056-50MG
APOSBICL4056-100MGEA 13530 CZK
APOSBICL4056-100MG APOSBICL4056-250MG APOSBICL4056-50MG
2-Chloro-4-nitrophenyl α-D-glucopyranoside ≥98%
2-Chloro-4-nitrophenyl α-D-glucopyranoside
Storage Temperature: Ambient
CAS číslo: 119047-14-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR