Tisk…

Oxcarbazepine-10-O-D-glucuronide ≥95%

Dodavatel: Apollo Scientific

BICL4053-1MG BICL4053-2MG
APOSBICL4053-1MGEA 13530 CZK
APOSBICL4053-1MG APOSBICL4053-2MG
Oxcarbazepine-10-O-D-glucuronide ≥95%
Oxcarbazepine-10-O-D-glucuronide
Storage Temperature: Ambient
CAS číslo: 144407-84-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR