6-O-Benzoyl-3-O-benzyl-1,2-O-isopropylidene-5-O-mesyl-α-D-glucofuranose ≥99%
Dodavatel: Apollo Scientific
BICL4045-1G
BICL4045-2G
BICL4045-5G
APOSBICL4045-1GEA
11180
CZK
APOSBICL4045-1G
APOSBICL4045-2G
APOSBICL4045-5G
6-O-Benzoyl-3-O-benzyl-1,2-O-isopropylidene-5-O-mesyl-α-D-glucofuranose ≥99%
6-O-Benzoyl-3-O-benzyl-1,2-O-isopropylidene-5-O-mesyl-α-D-glucofuranose
Storage Temperature:
Ambient |
CAS číslo:
134198-40-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...