Tisk…

3-O-Benzyl-D-glucopyranose ≥99%

Dodavatel: Apollo Scientific

BICL4043-1G BICL4043-2G BICL4043-5G
APOSBICL4043-1GEA 9560 CZK
APOSBICL4043-1G APOSBICL4043-2G APOSBICL4043-5G
3-O-Benzyl-D-glucopyranose ≥99%
3-O-Benzyl-D-glucopyranose
Storage Temperature: Ambient
CAS číslo: 10230-17-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR