Tisk…
(Z)-PUGNAc ≥99%
Dodavatel: Apollo Scientific
Synonyms: (Z)-O-(2-Acetamido-2-deoxy-D-glucopyranosylidene)amino N-phenylcarbamate, O-​(2-​Acetamido-​2-​deoxy-​D-​glucopyranosylidene)​amino Z-N-​phenyl carbamate
BICL4035-10MG BICL4035-25MG BICL4035-5MG
APOSBICL4035-10MGEA 7280 CZK
APOSBICL4035-10MG APOSBICL4035-25MG APOSBICL4035-5MG
(Z)-PUGNAc ≥99%
(Z)-PUGNAc
Vzorec: C₁₅H₁₉N₃O₇
MW: 353.33 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00145022
CAS číslo: 132489-69-1