Tisk…

2-Azidoethyl 2,3,4,6-tetra-O-acetyl-β-D-galactopyranoside ≥99%

Dodavatel: Apollo Scientific

BICL4031-1G BICL4031-2G BICL4031-500MG
APOSBICL4031-1GEA 14490 CZK
APOSBICL4031-1G APOSBICL4031-2G APOSBICL4031-500MG
2-Azidoethyl 2,3,4,6-tetra-O-acetyl-β-D-galactopyranoside ≥99%
2-Azidoethyl 2,3,4,6-tetra-O-acetyl-β-D-galactopyranoside
Storage Temperature: Ambient
CAS číslo: 139888-80-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR