Tisk…
1,6-Anhydro-β-D-cellotriose ≥98%
Dodavatel: Apollo Scientific
Synonyms: Cellotriosan
BICL4020-10MG BICL4020-25MG BICL4020-5MG
APOSBICL4020-10MGEA 16530 CZK
APOSBICL4020-10MG APOSBICL4020-25MG APOSBICL4020-5MG
1,6-Anhydro-β-D-cellotriose ≥98%
1,6-Anhydro-β-D-cellotriose
Storage Temperature: Ambient
CAS číslo: 78797-67-8