Tisk…
N-D-Glucopyranosyl-5-aminosalicylic acid ≥95%
Dodavatel: Apollo Scientific
Synonyms: 5-(D-Glucopyranosylamino)-2-hydroxybenzoic acid, N-D-Glucopyranosyl-5-ASA, Glucosyl-Mesalamine
BICL4006-1MG BICL4006-5MG BICL4006-2MG
APOSBICL4006-1MGEA 15940 CZK
APOSBICL4006-1MG APOSBICL4006-5MG APOSBICL4006-2MG
N-D-Glucopyranosyl-5-aminosalicylic acid ≥95%
N-D-Glucopyranosyl-5-aminosalicylic acid
Storage Temperature: Ambient
CAS číslo: 123135-21-7