Tisk…

Acetaminophen-O-β-D-glucuronide sodium salt ≥98%

Dodavatel: Apollo Scientific

Synonyms: Paracetamol glucuronide, Acetaminophen glucuronide, 4-Acetamidophenyl-O-β-D-glucuronide, p-Acetamidophenyl β-D-glucuronide sodium salt, Paracetamol-O-β-D-glucuronide, p-Acetylaminophenol-O-β-D-glucuronide

BICL4004-100MG BICL4004-25MG BICL4004-50MG
APOSBICL4004-100MGEA 22750 CZK
APOSBICL4004-100MG APOSBICL4004-25MG APOSBICL4004-50MG
Acetaminophen-O-β-D-glucuronide sodium salt ≥98%
Acetaminophen-O-β-D-glucuronide sodium salt
Vzorec: C₁₄H₁₆NO₈Na
Storage Temperature: Ambient
MDL Number: MFCD00153898
CAS číslo: 120595-80-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR