Acemetacin-acyl-β-D-glucuronide ≥98%
Dodavatel: Apollo Scientific
BICL4002-1MG
BICL4002-5MG
BICL4002-2MG
APOSBICL4002-1MGEA
11380
CZK
APOSBICL4002-1MG
APOSBICL4002-5MG
APOSBICL4002-2MG
Acemetacin-acyl-β-D-glucuronide ≥98%
Acemetacin-acyl-β-D-glucuronide
Storage Temperature:
Ambient |
CAS číslo:
1260603-31-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...