(R,S)-Ambrisentan-acyl-β-D-glucuronide ≥95%
Dodavatel: Apollo Scientific
BICL4001-1MG
BICL4001-5MG
BICL4001-2MG
APOSBICL4001-1MGEA
11380
CZK
APOSBICL4001-1MG
APOSBICL4001-5MG
APOSBICL4001-2MG
(R,S)-Ambrisentan-acyl-β-D-glucuronide ≥95%
(R,S)-Ambrisentan-acyl-β-D-glucuronide
Storage Temperature:
Ambient |
CAS číslo:
1106685-82-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...