Tisk…

Sesamolin

Dodavatel: Apollo Scientific
Varování

BIB6294-10MG BIB6294-1MG BIB6294-5MG
APOSBIB6294-10MGEA 11580 CZK
APOSBIB6294-10MG APOSBIB6294-1MG APOSBIB6294-5MG
Sesamolin
Sesamolin
Vzorec: C₂₀H₁₈O₇
Storage Temperature: Freezer
MDL Number: MFCD04112844
CAS číslo: 526-07-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR