Tisk…

4-Nitrophenyl-β-D-maltopyranoside

Dodavatel: Apollo Scientific
Varování

BIB6132-1G BIB6132-500MG
APOSBIB6132-1GEA 11580 CZK
APOSBIB6132-1G APOSBIB6132-500MG
4-Nitrophenyl-β-D-maltopyranoside
4-Nitrophenyl-β-D-maltopyranoside
Vzorec: C₁₈H₂₅NO₁₃
MW: 463,39 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00069844
CAS číslo: 56846-39-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR