Tisk…

5-Methoxy-L-tryptophan

Dodavatel: Apollo Scientific
Varování

Synonyms: (S)-2-Amino-3-(5-methoxy-1H-indol-3-yl)propanoic acid, H-Trp(5-OMe)-OH

BIB6103-1G BIB6103-250MG
APOSBIB6103-1GEA 7110 CZK
APOSBIB6103-1G APOSBIB6103-250MG
5-Methoxy-L-tryptophan
5-Methoxy-L-tryptophan
Vzorec: C₁₂H₁₄N₂O₃
MW: 234,251 g/mol
Bod tání: 253 °C
Storage Temperature: Freezer
MDL Number: MFCD01074513
CAS číslo: 25197-96-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR