Indole-3-acetyl-L-valine
Dodavatel: Apollo Scientific
|
|
Synonyms:
N-(3-Indolylacetyl)-L-valine
BIB6077-1G
BIB6077-500MG
APOSBIB6077-1GEA
5960
CZK
APOSBIB6077-1G
APOSBIB6077-500MG
Indole-3-acetyl-L-valine
Indole-3-acetyl-L-valine
Vzorec:
C₁₅H₁₈N₂O₃ MW: 274,315 g/mol Bod tání: 196…199 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00075402 CAS číslo: 57105-42-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...